Learn More
Invitrogen™ ZNF206 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5140341
Description
Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: PSPWPEESSR DQELAAVLEC LTFEDVPENK AWPAHPLGFG SRTPDKEEFK QEEPKGAAWP TPILAESQAD Highest antigen sequence indentity to the following orthologs: Mouse - 67%, Rat - 61%.
Specifications
| ZNF206 | |
| Polyclonal | |
| Unconjugated | |
| ZSCAN10 | |
| BC068284; Zfp206; zinc finger and SCAN domain containing 10; zinc finger and SCAN domain-containing protein 10; zinc finger and SCAN domains 10; zinc finger protein 206; zinc finger with KRAB and SCAN domains 10; Zkscan10; Znf206; ZSCAN10 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 84891 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.4 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q96SZ4 | |
| ZSCAN10 | |
| Recombinant protein corresponding to Human ZSCAN10. Recombinant protein control fragment (Product #RP-109592). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.