missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ZNF207 Polyclonal Antibody
GREENER_CHOICE

Product Code. 17151434 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17151434 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17151434 Supplier Invitrogen™ Supplier No. PA5113575

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: TSKLIHPDED ISLEERRAQL PKYQRNLPRP GQAPIGNPPV GPIGGMMPPQ For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 100%.

ZNF207 is a new candidate transcription factor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF207
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene ZNF207
Gene Accession No. O43670
Gene Alias 49 kDa zinc finger protein; 8430401D15Rik; BUB3-interacting and GLEBS motif-containing protein ZNF207; BUGZ; hBuGZ; Zep; Zfp207; Zinc finger protein 207; Znf207
Gene Symbols ZNF207
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human ZNF207.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7756
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.