missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF213 Monoclonal Antibody (OTI1A12), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA810318S
This item is not returnable.
View return policy
Description
ZNF213 Monoclonal antibody specifically detects ZNF213 in Human, Rat samples. It is validated for Western BlotSpecifications
| ZNF213 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q92844 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 10010, 252961 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI1A12 | |
| Unconjugated | |
| Tank | |
| CR53; putative transcription factor CR53; Zinc finger protein 213; zinc finger protein with KRAB and SCAN domains 21; ZKSCAN21; ZNF213; ZSCAN53 | |
| Full length human recombinant protein of human TANK produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human, Rat | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction