missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF217 Antibody (OTI3G5), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-74933CL1
This item is not returnable.
View return policy
Description
ZNF217 Monoclonal antibody specifically detects ZNF217 in Human samples. It is validated for Immunohistochemistry,Western Blot
Specifications
| ZNF217 | |
| Monoclonal | |
| CoraFluor 1 | |
| DKFZp781O2147, KIAA0335, MGC125707, MGC125710, zinc finger protein 518, zinc finger protein 518A, ZNF518 | |
| Human recombinant protein fragment corresponding to amino acids 730-1048 of human ZNF217(NP_006517) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Western Blot | |
| OTI3G5 | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| 7764 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction