missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF217 Antibody, PerCP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-78188PCP
This item is not returnable.
View return policy
Description
ZNF217 Polyclonal antibody specifically detects ZNF217 in Human samples. It is validated for Immunoprecipitation, Western Blot (Negative)
Specifications
| ZNF217 | |
| Polyclonal | |
| Immunoprecipitation, Western Blot (Negative) | |
| DKFZp781O2147, KIAA0335, MGC125707, MGC125710, zinc finger protein 518, zinc finger protein 518A, ZNF518 | |
| The immunogen for this product maps to a region between residue 100 and 150 of human Zinc Finger Protein 217 using the numbering given in entry NP_006517.1 (GeneID 7764). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Immunoprecipitation | |
| PerCP | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 7764 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction