missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF217 Mouse anti-Human, Clone: OTI2B4, liquid, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies TA809052
This item is not returnable.
View return policy
Description
Binds to the promoters of target genes and functions as repressor. Promotes cell proliferation and antagonizes cell death. Promotes phosphorylation of AKT1 at ′Ser-473′.Specifications
| ZNF217 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| O75362 | |
| ZNF217 | |
| Human recombit protein fragment corresponding to amino acids 730-1048 of human ZNF217 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI2B4 | |
| Unconjugated | |
| ZNF217 | |
| ZABC1 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 7764 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction