Learn More
ZNF217 Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 24869-1-AP-150UL
 View more versions of this product
Product Code. 18055405
  • € 410.00 / 150µL
Description
The ZNF217 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human ZNF217. This antibody recognizes human antigen. The ZNF217 antibody has been validated for the following applications: IP, WB, ELISA analysis.
Specifications
Specifications
ZNF217
350 μg/mL
Rabbit
Primary
Human
150 kDa
IgG
Western Blot, Immunoprecipitation, ELISA
ZABC1, ZFP217, zinc finger protein 217, ZNF217
150 μL
7764
Store at -20°C. Stable for one year after shipment.