missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF224 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA5116266
Additional Details : Weight : 0.01000kg
Description
Matrix-assisted laser desorption ionization-time of flight analysis and examination of protein profiles from the SwissProt database revealed that the previously defined p97 repressor is ZNF224, a zinc finger protein. ZNF224, a Kruppel-like zinc finger transcription factor, is the repressor protein that specifically binds to the negative cis-element AldA-NRE and affects the AldA-NRE-mediated transcription.Specifications
ZNF224 | |
Polyclonal | |
PBS with 0.09% sodium azide | |
Q9NZL3 | |
ZNF224 | |
KLH conjugated synthetic peptide between 127-155 amino acids from the N-terminal region of human ZNF224 | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
ZNF224 | |
BMZF2; BMZF-2; Bone marrow zinc finger 2; KOX22; Zinc finger protein 224; zinc finger protein 233; Zinc finger protein 255; zinc finger protein 27; Zinc finger protein KOX22; ZNF224; ZNF233; ZNF255; ZNF27 | |
Rabbit | |
Protein A | |
RUO | |
7767 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |