missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ZNF227 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15584722
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15584722 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15584722 Supplier Invitrogen™ Supplier No. PA541247

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Peptide sequence: ESGDCFNKSS FHSYQSNHTG EKSYRCDSCG KGFSSSTGLI IHYRTHTGEK Sequence homology: Human: 100%.

ZNF227 is a new candidate transcription factor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF227
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene ZNF227
Gene Accession No. Q86WZ6
Gene Alias Zinc finger protein 227; ZNF227
Gene Symbols ZNF227
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human ZNF227.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7770
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.