missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF230 Antibody (OTI1E2), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74934B
This item is not returnable.
View return policy
Description
ZNF230 Monoclonal specifically detects ZNF230 in Human samples. It is validated for Immunohistochemistry.
Specifications
| ZNF230 | |
| Monoclonal | |
| Biotin | |
| PBS with 0.05% Sodium Azide | |
| ZNF230 | |
| Full length recombinant protein of human ZNF230 (NP_006291) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG1 |
| Immunohistochemistry | |
| OTI1E2 | |
| Immunohistochemistry | |
| FLJ12515, FLJ14855, zinc finger protein 502 | |
| Mouse | |
| Affinity Purified | |
| Zinc Finger | |
| 7773.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction