Learn More
Invitrogen™ ZNF230 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA564906
Description
Immunogen sequence: GKTIAEAGPHE DCPCQQIWEQ TASDLTQSQD SIINNSHFFE QGDVPSQVEA GLSIIHTGQK PSQNGKCKQS FSDVAIFDPP QQFHSGEKS Highest antigen sequence identity to the following orthologs - mouse 29%, rat 29%.
ZNF230 is predicted to enable DNA-binding transcription factor activity, RNA polymerase II-specific and RNA polymerase II cis-regulatory region sequence-specific DNA binding activity. This gene is predicted to be active in the nucleus and is predicted to be involved in regulation of transcription by RNA polymerase II. ZN230 may be involved in transcriptional regulation.
Specifications
| ZNF230 | |
| Polyclonal | |
| Unconjugated | |
| ZNF230 | |
| 2610110L04Rik; AA792898; AU022812; C3HC4-like zinc finger protein; FDZF2; MGC8715; RING finger protein 141; Rnf141; RNF141 protein; ZFP26; ZFP36; Zinc finger protein 230; Zinc Finger Protein FDZF2; ZNF230 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 7773 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9UIE0 | |
| ZNF230 | |
| Recombinant Human ZNF230. Recombinant protein control fragment (Product #RP-104551). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.