missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF232 Antibody (PCRP-ZNF232-1D5), Novus Biologicals™
Mouse Monoclonal Antibody
€ 232.00 - € 462.00
Specifications
| Antigen | ZNF232 |
|---|---|
| Clone | PCRP-ZNF232-1D5 |
| Concentration | 0.2 mg/mL |
| Dilution | Flow Cytometry : 1-2 μg/million cells, Immunohistochemistry-Paraffin : 1-2 μg/mL |
| Applications | Flow Cytometry, Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18671494
|
Novus Biologicals
NBP3-23599-20ug |
20 μg |
€ 232.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18639174
|
Novus Biologicals
NBP3-23599-100ug |
100 μg |
€ 462.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ZNF232 Monoclonal antibody specifically detects ZNF232 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry (Paraffin)Specifications
| ZNF232 | |
| 0.2 mg/mL | |
| Flow Cytometry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Human | |
| zinc finger protein 500, Zinc finger protein with KRAB and SCAN domains 18, ZKSCAN18KIAA0557 | |
| Recombinant fragment (around aa29-109) of human ZNF232 protein | |
| Primary | |
| Store at 4°C. |
| PCRP-ZNF232-1D5 | |
| Flow Cytometry : 1-2 μg/million cells, Immunohistochemistry-Paraffin : 1-2 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| 10 mM PBS with 0.05% BSA | |
| 7775 | |
| IgG2b | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title