missing translation for 'onlineSavingsMsg'
Learn More

ZNF232 Antibody (PCRP-ZNF232-2B3) [Janelia Fluor™ 525], Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals (Bio-Techne) NBP3-24043JF525

Product Code. 30163193

  • € 648.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

ZNF232 Monoclonal antibody specifically detects ZNF232 in Human samples. It is validated for Flow Cytometry,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ZNF232
Monoclonal
Janelia Fluor 525
zinc finger protein 500, Zinc finger protein with KRAB and SCAN domains 18, ZKSCAN18KIAA0557
Recombinant fragment (around aa29-109) of human ZNF232 protein
0.1 mL
Primary
Human
Purified
Flow Cytometry, Immunohistochemistry (Paraffin)
PCRP-ZNF232-2B3
50mM Sodium Borate
Mouse
Protein A or G purified
RUO
7775
Store at 4°C in the dark.
IgG2a
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.