missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ZNF234 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15799570 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15799570 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15799570 Supplier Invitrogen™ Supplier No. PA551621

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: DPVQRNLYQD VMLENFRNLL SVGHHPFKHD VFLLEKEKKL DIMKTATQRK GKSADKIQSE VETVPEAGRH EELYWGQIWK QIASDLIKYE DSMISISRFP RQGDLSCQVR AGLYTTHTGQ K Highest antigen sequence identity to the following orthologs: Mouse - 38%, Rat - 36%.

May be involved in transcriptional regulation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF234
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZNF234
Gene Accession No. Q14588
Gene Alias C2-H2 type zinc finger protein; Homo sapiens zinc finger 234; HZF4; zinc finger protein 234; zinc finger protein 234, partial sequence; zinc finger protein 269; zinc finger protein 4; zinc finger protein HZF4; ZNF234; ZNF269
Gene Symbols ZNF234
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ZNF234. Recombinant protein control fragment (Product #RP-99721).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10780
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.