missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZNF238 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5112797
This item is not returnable.
View return policy
Description
ZNF238 Polyclonal Antibody for Western Blot, IHC (P), ELISA
ZNF238 is the sequence-specific DNA-binding protein with transcriptional repression activity. ZNF238 binds the consensus DNA sequence 5'-[AC]ACATCTG[GT][AC]-3' which contains the E box core. ZNF238 may play a role in the organization of chromosomes in the nucleus.C2H2-type zinc finger proteins, such as ZNF238, act on the molecular level as transcriptional activators or repressors and are involved in chromatin assembly.
Specifications
| ZNF238 | |
| Polyclonal | |
| Unconjugated | |
| ZBTB18 | |
| 58 kDa repressor protein; BTB/POZ domain protein ZF238; C2H2-171; MRD22; novel zinc finger domain containing protein; RP58; rRP58; si:ch211-221n23.1; TAZ1; TAZ-1; Transcriptional repressor RP58; transcriptional represssor; translin-associated zinc finger protein 1; Zbtb18; Zfp238; Zfp-238; zinc finger and BTB domain containing 18; zinc finger and BTB domain-containing protein 18; Zinc finger protein 238; zinc finger protein C2H2-171; Znf238 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 10472 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 3.5 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q99592 | |
| ZBTB18 | |
| Recombinant Human Zinc finger and BTB domain-containing protein 18 protein (62-364AA). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction