missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ZNF238 Polyclonal Antibody
GREENER_CHOICE

Product Code. 17884780
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17884780 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17884780 Supplier Invitrogen™ Supplier No. PA5140267

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: EDASSCSDKV ESLSDGSSHI AGDLPSDEDE GEDEKLNILP SKRDLAAEPG NMWMRLPSDS AGI Highest antigen sequence indentity to the following orthologs: Mouse - 97%, Rat - 95%.

ZNF238 is the sequence-specific DNA-binding protein with transcriptional repression activity. ZNF238 binds the consensus DNA sequence 5'-[AC]ACATCTG[GT][AC]-3' which contains the E box core. ZNF238 may play a role in the organization of chromosomes in the nucleus.C2H2-type zinc finger proteins, such as ZNF238, act on the molecular level as transcriptional activators or repressors and are involved in chromatin assembly.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF238
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZBTB18
Gene Accession No. Q99592
Gene Alias 58 kDa repressor protein; BTB/POZ domain protein ZF238; C2H2-171; MRD22; novel zinc finger domain containing protein; RP58; rRP58; si:ch211-221n23.1; TAZ1; TAZ-1; Transcriptional repressor RP58; transcriptional represssor; translin-associated zinc finger protein 1; Zbtb18; Zfp238; Zfp-238; zinc finger and BTB domain containing 18; zinc finger and BTB domain-containing protein 18; Zinc finger protein 238; zinc finger protein C2H2-171; Znf238
Gene Symbols ZBTB18
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ZBTB18. Recombinant protein control fragment (Product #RP-109382).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10472
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.