Learn More
ZNF24 Antibody, mFluor Violet 450 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97861MFV450
This item is not returnable.
View return policy
Description
ZNF24 Polyclonal antibody specifically detects ZNF24 in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications
| Produced in rabbits immunized with E. coli-derived Human ZNF24 fragment. | |
| Store at 4C in the dark. |