Learn More
ZNF24 Antibody, mFluor Violet 450 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP2-97861MFV450
Product Code. 18502104
  • € 428.00 / 0.10mL
Description
ZNF24 Polyclonal antibody specifically detects ZNF24 in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Produced in rabbits immunized with E. coli-derived Human ZNF24 fragment.
Store at 4C in the dark.