missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZNF24 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA582865
This item is not returnable.
View return policy
Description
Immunogen sequence: LESELDDPGQ PVSLRRRKRE VLVEDMVSQE EAQGLPSSEL DAVENQLKWA SWELHSLRHC DDDGRTENGA LAPKQELPSA LESHEVPGTL SMGVPQIFKY GETCFPKGRF ERKRNPSRKK QHICDECGKH FS Highest antigen sequence indentity to the following orthologs: Rat - 91%, Mouse - 89%.
Transcription factor required for myelination of differentiated oligodendrocytes. Required for the conversion of oligodendrocytes from the premyelinating to the myelinating state. In the developing central nervous system (CNS), involved in the maintenance in the progenitor stage by promoting the cell cycle. Specifically binds to the 5'-TCAT-3' DNA sequence. Has transcription repressor activity in vitro.
Specifications
| ZNF24 | |
| Polyclonal | |
| Unconjugated | |
| Znf24 | |
| 3526401F17Rik; 5033419P20Rik; AI480505; AI642085; AL024422; Hmcns; Hypomyelinated CNS protein; KOX17; LOW QUALITY PROTEIN: zinc finger protein 24; retinoic acid suppression protein A; RSG-A; ZF-12; Zfp191; zfp-191; Zfp24; Zinc finger and SCAN domain-containing protein 3; zinc finger protein 191; Zinc finger protein 24; zinc finger protein 24 (KOX 17); zinc finger protein 24-like protein; zinc finger protein KOX17; Zinc finger protein ZF-12; zinc finger ZF-12; Znf191; Znf24; Znp191; ZSCAN3 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 7572 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P17028 | |
| Znf24 | |
| Recombinant protein corresponding to Human ZNF24. Recombinant protein control fragment (Product #RP-100678). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction