missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ZNF24 Polyclonal Antibody
GREENER_CHOICE

Product Code. 16521082
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16521082 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16521082 Supplier Invitrogen™ Supplier No. PA598254

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

ZNF24 Polyclonal Antibody for Western Blot, IHC (P), ELISA

Transcription factor required for myelination of differentiated oligodendrocytes. Required for the conversion of oligodendrocytes from the premyelinating to the myelinating state. In the developing central nervous system (CNS), involved in the maintenance in the progenitor stage by promoting the cell cycle. Specifically binds to the 5'-TCAT-3' DNA sequence. Has transcription repressor activity in vitro.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF24
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.39 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Znf24
Gene Accession No. P17028
Gene Alias 3526401F17Rik; 5033419P20Rik; AI480505; AI642085; AL024422; Hmcns; Hypomyelinated CNS protein; KOX17; LOW QUALITY PROTEIN: zinc finger protein 24; retinoic acid suppression protein A; RSG-A; ZF-12; Zfp191; zfp-191; Zfp24; Zinc finger and SCAN domain-containing protein 3; zinc finger protein 191; Zinc finger protein 24; zinc finger protein 24 (KOX 17); zinc finger protein 24-like protein; zinc finger protein KOX17; Zinc finger protein ZF-12; zinc finger ZF-12; Znf191; Znf24; Znp191; ZSCAN3
Gene Symbols Znf24
Host Species Rabbit
Immunogen Recombinant Human Zinc finger protein 24 protein (169-368AA).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7572
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.