missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ZNF258 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA567211

Product Code. 15919884

  • € 578.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: VGFIICPGSKE SSPRPQCVIC GEILSSENMK PANLSHHLKT KHSELENKPV DFFEQKSLEM ECQNSSLKKC LLVEKSLVKA SYLIAFQTAA S Highest antigen sequence identity to the following orthologs - mouse 80%, rat 82%.

ZNF258 is part of a novel, putative, zinc-binding motif (MYM) family which encodes proteins that maintain the repeats of the MYM motif.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ZNF258
Polyclonal
Unconjugated
ZMYM6
Buster2; KIAA1353; MYM; transposase-like protein; transposon-derived Buster2 transposase-like protein; ZBED7; zinc finger MYM-type containing 6; zinc finger MYM-type protein 6; Zinc finger protein 258; zinc finger, BED-type containing 7; zinc finger, MYM-type 6; zinc finger, MYM-type containing 6; ZMYM6; ZNF198L4; ZNF258
Rabbit
Antigen affinity chromatography
RUO
9204
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunocytochemistry
0.05 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
O95789
ZMYM6
Recombinant Human ZNF258. Recombinant protein control fragment (Product #RP-107845).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.