missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZNF260 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA555981
This item is not returnable.
View return policy
Description
Immunogen sequence: MIGMLESLQH ESDLLQHDQI HTGEKPYECN ECRKTFSLKQ NLVEHKKMHT GEKSH Highest antigen sequence identity to the following orthologs: Mouse - 65%, Rat - 67%.
Transcription factor that acts as a cardiac regulator and an effector of alpha1-adrenergic signaling. Binds to PE response elements (PERE) present in the promoter of genes such as ANF/NPPA and acts as a direct transcriptional activator of NPPA. Also acts as a cofactor with GATA4, a key cardiac regulator.
Specifications
| ZNF260 | |
| Polyclonal | |
| Unconjugated | |
| ZNF260 | |
| AI480997; Ozrf1; PEX1; Zfp260; zfp-260; Zfp63; Zinc finger protein 260; Znf260 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 339324 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ChIP Assay, Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q3ZCT1 | |
| ZNF260 | |
| Recombinant protein corresponding to Human ZNF260. Recombinant protein control fragment (Product #RP-95537). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction