missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ZNF263 Polyclonal Antibody

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA548786

Product Code. 15623085

  • € 530.00 / 400µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

ZNF263 Polyclonal Antibody for Western Blot

Might play an important role in basic cellular processes as a transcriptional repressor.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ZNF263
Polyclonal
PBS with 0.09% sodium azide; pH 7.4
O14978
ZNF263
KLH conjugated synthetic peptide between 318-346 amino acids from the Central region of human ZNF263.
400 μL
Primary
Human
Antibody
IgG
Western Blot
Unconjugated
ZNF263
FPM315; zinc finger protein 263; Zinc finger protein FPM315; zinc finger protein with KRAB and SCAN domains 12; ZKSCAN12; ZNF263; ZSCAN44
Rabbit
Antigen affinity chromatography, Protein A
RUO
10127
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.