missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZNF264 Monoclonal Antibody (OTI8F4)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA527309
This item is not returnable.
View return policy
Description
ZNF264 Monoclonal Antibody for Western Blot
ZNF264 gene encodes a zinc finger protein and belongs to the krueppel C2H2-type zinc-finger protein family. Zinc finger proteins are often localized in the nucleus, bind nucleic acids, and regulate transcription.
Specifications
| ZNF264 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O43296 | |
| ZNF264 | |
| Full length human recombinant protein of ZNF264 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI8F4 | |
| Unconjugated | |
| ZNF264 | |
| KIAA0412; Zinc finger protein 264; ZNF264 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 9422 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction