Learn More
ZNF264 Monoclonal Antibody (OTI8G5), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA810500S
This item is not returnable.
View return policy
Description
ZNF264 Monoclonal antibody specifically detects ZNF264 in Human samples. It is validated for Western BlotSpecifications
| ZNF264 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q86UA6 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 84268 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI8G5 | |
| Unconjugated | |
| RPAIN | |
| KIAA0412; Zinc finger protein 264; ZNF264 | |
| Full length human recombinant protein of human RPAIN produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |