missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF264 Monoclonal Antibody (OTI8E9), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA810501
This item is not returnable.
View return policy
Description
ZNF264 gene encodes a zinc finger protein and belongs to the krueppel C2H2-type zinc-finger protein family. Zinc finger proteins are often localized in the nucleus, bind nucleic acids, and regulate transcription.Specifications
| ZNF264 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| O43296 | |
| ZNF264 | |
| Full length human recombit protein of human ZNF264 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI8E9 | |
| Unconjugated | |
| ZNF264 | |
| zinc finger protein 264 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 9422 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction