Learn More
Invitrogen™ ZNF264 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA568963
Product Code. 15867373
  • € 520.00 / 100µL
Description
This target displays homology in the following species: Cow: 86%; Dog: 86%; Horse: 93%; Human: 100%; Mouse: 86%; Pig: 92%; Rat: 83%.
ZNF264 gene encodes a zinc finger protein and belongs to the krueppel C2H2-type zinc-finger protein family. Zinc finger proteins are often localized in the nucleus, bind nucleic acids, and regulate transcription.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
ZNF264
Polyclonal
Unconjugated
ZNF264
KIAA0412; Zinc finger protein 264; ZNF264
Rabbit
Affinity Chromatography
RUO
9422
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Western Blot, Immunocytochemistry
0.5 mg/mL
PBS with 2% sucrose and 0.09% sodium azide
O43296
ZNF264
synthetic peptide directed towards the N terminal region of human ZNF264.