missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZNF266 Monoclonal Antibody (4G5)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA521156
This item is not returnable.
View return policy
Description
Peptide Sequence: GRAFTVSSCL SQHMKIHVGE KPYECKECGI AFTRSSQLTE HLKTHTAKDP FECKICGKSF RNSSCLSDHF RIH.
This gene encodes a protein containing many tandem zinc-finger motifs. Zinc fingers are protein or nucleic acid-binding domains, and may be involved in a variety of functions, including regulation of transcription. This gene is located in a cluster of similar genes encoding zinc finger proteins on chromosome 19. Alternative splicing results in multiple transcript variants for this gene.
Specifications
| ZNF266 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative; pH 7.4 | |
| Q14584 | |
| ZNF266 | |
| ZNF266 (NP_006622.2, 274 a.a. ~ 346 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a κ |
| ELISA, Immunohistochemistry (PFA fixed), Western Blot | |
| 4G5 | |
| Unconjugated | |
| ZNF266 | |
| HZF1; KIAA2007; zinc finger protein 1; zinc finger protein 266; Zinc finger protein HZF1; ZNF266 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 10781 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction