missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ZNF268 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA588851

Product Code. 16536941

  • € 530.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: MATRVRTASI WVPPLQERNS SWDRIRKLQG QESILGQGTP GLQPLPGTPR QKQKSRRIEK VLEWLFISQE QPKITKSWGP LSFMDVFVDF TWEEWQLLDP AQKCLYRSVM LENYSNLVSL GYQHTKPDII FKLEQGEELC MVQAQVPNQT CPNTVWKIDD LMDWHQENKD KLGSTAKSFE CTTFGKLCLL STKYLSRQKP HKCGTHGKSL KYIDFTSDYA RNNPNGFQVH GKSFFHSKHE QTVIGIKYCE SIESGKTVNK KSQLMCQQMY; Positive Samples: Mouse kidney, Rat ovary; Cellular Location: Cytoplasm, Nucleus, Nucleus.

Zinc-finger proteins contain DNA-binding domains and have a wide variety of functions, most of which encompass some form of transcriptional activation or repression. The majority of zinc-finger proteins contain a krueppel-type DNA binding domain and a KRAB domain, which is thought to interact with KAP1, thereby recruiting histone modifying proteins. Zinc finger protein 268 (ZNF268), also known as zinc finger protein 3 or HZF3, is a 947 amino acid protein belonging to the krueppel C2H2-type zinc-finger protein family. ZNF268 contains 24 C2H2-type zinc fingers and one KRAB domain. Localized to the nucleus, ZNF268 is involved in transcriptional regulation and is highly expressed in three to five week old embryos. ZNF268 has been implicated in human leukemia, due to the identification of an alternatively spliced form in leukemia patients. Two named isoforms of ZNF268 exist as a result of alternative splicing events.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ZNF268
Polyclonal
Unconjugated
ZNF268
HZF3; zinc finger protein 268; zinc finger protein 3; zinc finger protein HZF3; ZNF268
Rabbit
Affinity Chromatography
RUO
10795
-20°C, Avoid Freeze/Thaw Cycles
Liquid
ELISA, Western Blot
1.28 mg/mL
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3
Q14587
ZNF268
Recombinant fusion protein containing a sequence corresponding to amino acids 1-270 of human ZNF268 (NP_0011593531).
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.