missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF276 Antibody, DyLight 594, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-03342DL594
This item is not returnable.
View return policy
Description
ZNF276 Polyclonal antibody specifically detects ZNF276 in Human, Mouse samples. It is validated for Western Blot
Specifications
| ZNF276 | |
| Polyclonal | |
| Western Blot | |
| DNA-binding protein, KIAA1710Zfp46, zinc finger protein 436, ZNF | |
| The immunogen recognized by this antibody maps to a region between residue 564 and 614 of human zinc finger protein 276 using the numbering given in entry NP_001106997.1 (GeneID 92822). | |
| 0.1 mL | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot | |
| DyLight 594 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 92822 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction