missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF280A Monoclonal Antibody (OTI2H3), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA800173S
This item is not returnable.
View return policy
Description
ZNF280A Monoclonal antibody specifically detects ZNF280A in Human samples. It is validated for Flow Cytometry, Western BlotSpecifications
| ZNF280A | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P12757 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 6498 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Flow Cytometry, Western Blot | |
| OTI2H3 | |
| Unconjugated | |
| SKIL | |
| 3'OY11.1; SUHW1; Suppressor of hairy wing homolog 1; zinc finger protein 280; zinc finger protein 280A; Zinc finger protein 636; ZNF280; ZNF280A; ZNF636 | |
| Human recombinant protein fragment corresponding to amino acids 307-684 of human SKIL produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction