missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF281 Antibody, Biotin, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-76551B
This item is not returnable.
View return policy
Description
ZNF281 Polyclonal antibody specifically detects ZNF281 in Human, Mouse samples. It is validated for Western Blot, ELISA
Specifications
| ZNF281 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| 23528 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot, ELISA | |
| Biotin | |
| FLJ20557, zinc finger protein 416 | |
| Antibody was raised against a 19 amino acid synthetic peptide near the carboxy terminus of human ZFP281. The immunogen is located within amino acids 600 - 650 of ZFP281. Amino Acid Squence: YSNKSESQKEDPFNIAEPR | |
| 0.1 mL | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction