missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF281 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 280.00 - € 556.50
Specifications
| Antigen | ZNF281 |
|---|---|
| Dilution | Western Blot 0.04-0.4 ug/ml, Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml |
| Applications | Western Blot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18452402
|
Novus Biologicals
NBP2-13564-25ul |
25ul |
€ 280.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18077499
|
Novus Biologicals
NBP2-13564 |
0.1 mL |
€ 589.00 € 556.50 / 0.10mL Save € 32.50 5% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ZNF281 Polyclonal specifically detects ZNF281 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
| ZNF281 | |
| Western Blot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 23528 | |
| This antibody was developed against a recombinant protein corresponding to the amino acids: SSPLHNHTLFPEKQIYTTSPLECGFGQSVTSVLPSSLPKPPFGMLFGSQPGLYLSALDATHQQLTPSQELDDLIDSQKNLETSSAFQSSSQKLTSQKEQ | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot 0.04-0.4 ug/ml, Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml | |
| Polyclonal | |
| Rabbit | |
| Human | |
| FLJ20557, zinc finger protein 416 | |
| ZNF281 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title