Learn More
ZNF281 Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 24088-1-AP-150UL
 View more versions of this product
Product Code. 18065565
  • € 410.00 / 150µL
Description
The ZNF281 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human ZNF281. This antibody recognizes human antigen. The ZNF281 antibody has been validated for the following applications: IF, WB, ELISA analysis.
Specifications
Specifications
ZNF281
200 μg/mL
Rabbit
Primary
Human
150 kDa
IgG
Western Blot, Immunofluorescence, ELISA
GZP1, Transcription factor ZBP 99, ZBP 99, ZBP99, zinc finger protein 281, ZNF281, ZNP 99
150 μL
23528
Store at -20°C. Stable for one year after shipment.