missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF286A Monoclonal Antibody (OTI7D2), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA803480S
This item is not returnable.
View return policy
Description
ZNF286A Monoclonal antibody specifically detects ZNF286A in Human samples. It is validated for Western BlotSpecifications
| ZNF286A | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9H7C4 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 81493 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI7D2 | |
| Unconjugated | |
| SYNC | |
| KIAA1874; Zinc finger protein 286A; zinc finger protein 624; zinc finger protein ZNF286; ZNF286; ZNF286A | |
| Full length human recombinant protein of human SYNC produced in SF9 cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction