missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ZNF287 Polyclonal Antibody
GREENER_CHOICE

Product Code. 16617744
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16617744 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16617744 Supplier Invitrogen™ Supplier No. PA5101262

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total ZNF287.

This gene encodes a member of the krueppel family of zinc finger proteins, suggesting a role as a transcription factor. Its specific function has not been determined. This gene is located near the Smith-Magenis syndrome region on chromosome 17.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF287
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene ZNF287
Gene Accession No. Q9EQB9, Q9HBT7
Gene Alias B230333C16Rik; mszf16; mszf74; Skat2; SKAT-2; Zfp287; Zfp-287; Zinc finger protein 287; zinc finger protein SKAT2; zinc finger protein SKAT-2; zinc finger protein with KRAB and SCAN domains 13; ZKSCAN13; Znf287; ZSCAN45
Gene Symbols Zfp287, ZNF287
Host Species Rabbit
Immunogen A synthesized peptide derived from human ZNF287(Accession Q9HBT7), corresponding to amino acid residues E244-S294.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 170740, 57336
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.