missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ZNF287 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA583078

Product Code. 16370035

  • € 578.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: PTVIPILEEP WMVIKEILEG PSPEWETKAQ ACTPVEDMSK LTKEETHTIK LEDSYDYDDR LERRGKGGFW KIHTDERGFS LKSVLSQEYD PTEECLSKYD IYRNNFEKHS NLIVQFDTQL DNKTSVYNEG RA Highest antigen sequence indentity to the following orthologs: Mouse - 63%, Rat - 71%.

This gene encodes a member of the krueppel family of zinc finger proteins, suggesting a role as a transcription factor. Its specific function has not been determined. This gene is located near the Smith-Magenis syndrome region on chromosome 17.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ZNF287
Polyclonal
Unconjugated
ZNF287
B230333C16Rik; mszf16; mszf74; Skat2; SKAT-2; Zfp287; Zfp-287; Zinc finger protein 287; zinc finger protein SKAT2; zinc finger protein SKAT-2; zinc finger protein with KRAB and SCAN domains 13; ZKSCAN13; Znf287; ZSCAN45
Rabbit
Antigen affinity chromatography
RUO
57336
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
0.2 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q9HBT7
ZNF287
Recombinant protein corresponding to Human ZNF287. Recombinant protein control fragment (Product #RP-99746).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.