missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ ZNF295 Antibody [PerCP], Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NB100-74655PCP
This item is not returnable.
View return policy
Description
ZNF295 Polyclonal antibody specifically detects ZNF295 in Human samples. It is validated for Immunohistochemistry,Immunoprecipitation,Immunohistochemistry (Paraffin),Western Blot (Negative),Immunocytochemistry/ Immunofluorescence
Specifications
| ZNF295 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 49854 | |
| Store at 4°C in the dark. | |
| IgG |
| Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Western Blot (Negative), Immunocytochemistry/Immunofluorescence | |
| PerCP | |
| MGC15716, Zfp414, zinc finger protein 414 | |
| The immunogen recognized by this antibody maps to a region between residue 875 and 925 of human zinc finger protein 295 using the numbering given in entry NP_065778.3 (GeneID 49854). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction