Learn More
Invitrogen™ ZNF295 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA555141
Description
Immunogen sequence: QPEPNKVNHI VTTKDDNVFS DSSEQVNFDS EDSSCLPEDL SLSKQLKIQV KEEPVEEAEE EAPEASTAPK EAGPSKEASL WPCEKCGKMF TVHKQLERHQ ELLCSVKPFI CHVCNKAFRT NFRLWSHFQS HMSQASEESA HKESEVCP Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 86%.
ZNF295 contains 1 BTB (POZ) domain and 8 C2H2-type zinc fingers. ZNF295 may be involved in transcriptional regulation.
Specifications
| ZNF295 | |
| Polyclonal | |
| Unconjugated | |
| ZBTB21 | |
| 5430437K12Rik; B430213I24Rik; KIAA1227; mKIAA1227; ZBTB21; Zfp295; zinc finger and BTB domain containing 21; zinc finger and BTB domain-containing protein 21; Zinc finger protein 295; ZNF295 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 49854 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9ULJ3 | |
| ZBTB21 | |
| Recombinant protein corresponding to Human ZNF295. Recombinant protein control fragment (Product #RP-100680). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.