missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ZNF306 Polyclonal Antibody
GREENER_CHOICE

Product Code. 13229918
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
13229918 100 μL 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13229918 Supplier Invitrogen™ Supplier No. PA521718

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Recommended positive controls: K562. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

ZNF306 is a protein coding gene. Transcriptional factor that binds to the consensus sequence 5'-[GT][AG][AGT]GGGG-3' and acts as a repressor of autophagy. Specifically represses expression of genes involved in autophagy and lysosome biogenesis/function such as MAP1LC3B, ULK1 or WIPI2. Associates with chromatin at the ITGB4 and VEGF promoters. Also acts as a transcription activator and promotes cancer cell progression and/or migration in various tumors and myelomas.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF306
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.025% ProClin 300; pH 7
Gene ZKSCAN3
Gene Accession No. Q9BRR0
Gene Alias 2810435N07Rik; AA408594; AI132359; dJ874C20.1; dJ874C20.1.; mszf35; SCAN-KRAB-zinc finger gene 1; SCAN-KRAB-zinc finger protein; Skz1; Zf47; ZFP306; Zfp307; Zfp47; zfp-47; Zinc finger and SCAN domain-containing protein 13; zinc finger protein 306; Zinc finger protein 307; zinc finger protein 309; Zinc finger protein 47 homolog; zinc finger protein with KRAB and SCAN domains 3; zinc finger protein zfp47; zinc finger with KRAB and SCAN domains 3; Zkscan3; ZNF306; ZNF309; ZSCAN13; ZSCAN35
Gene Symbols ZKSCAN3
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human ZKSCAN3. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80317
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.