missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF307 Antibody (OTI6G6), Alexa Fluor™ 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74936AF350
This item is not returnable.
View return policy
Description
ZNF307 Monoclonal specifically detects ZNF307 in Human samples. It is validated for Western Blot.
Specifications
| ZNF307 | |
| Monoclonal | |
| Alexa Fluor 350 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| ZKSCAN4 | |
| Recombinant protein fragment corresponding to amino acids 1-316 of human ZNF307 (NP_061983) produced in E.coli. | |
| Primary | |
| Human | |
| IgG1 |
| Western Blot | |
| OTI6G6 | |
| Western Blot | |
| EC 2.1.1.43, EC 4.2.1.52, FLJ32136, P1P373C6, p373c6.1, Zinc finger protein 307p373c6.1 (novel C2H2 type zinc finger protein), Zinc finger protein 427zinc finger protein with KRAB and SCAN domains 4, zinc finger with KRAB and SCAN domains 4, ZNF307P373c6.1, ZNF427 | |
| Mouse | |
| 0.1 mL | |
| 387032 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction