missing translation for 'onlineSavingsMsg'
Learn More

ZNF311 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

€ 145.00 - € 410.00

Specifications

Antigen ZNF311
Concentration 0.41 mg/mL
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16842255
View Documents
Proteintech
23243-1-AP-20UL
20 μL
€ 145.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16832255
View Documents
Proteintech
23243-1-AP-150UL
150 μL
€ 410.00
150µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This antibody specifically reacts with ZNF311 protein.

ZNF311 gene ontology annotations related to this gene include regulation of transcription, DNA-templated.
Specifications

Specifications

ZNF311
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
Rabbit
Human
Q5JNZ3
282890
ZNF311 Fusion Protein Ag19624
Primary
-20°C
ZNF311
0.41 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
zf31, ZFP31, zinc finger protein 311, Zinc finger protein zfp 31, ZNF311
ZNF311
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.