missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF312 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94378-0.02ml
This item is not returnable.
View return policy
Description
ZNF312 Polyclonal antibody specifically detects ZNF312 in Human, Rat samples. It is validated for Western Blot
Specifications
| ZNF312 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| FEZ family zinc finger 2, fez family zinc finger protein 2, FEZLFEZ, FKSG36, FLJ10142, Forebrain embryonic zinc finger-like protein 2, Zfp312, Zinc finger protein 312TOFZNF312ZFP312, Zinc finger protein Fez-like | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-280 of human FEZF2 (NP_060478.3). MASSASLETMVPPACPRAGASPATSKTLAFSIERIMAKTSEPRAPFEPRPGALEADGSQGKKLLNLCSPLPCMIPLQPLGYEVPSKTLLSYSELWKSSLRAGGGGGGGGGGGGGGGGAPVCGASGLCKTNCGVCCKAELGLAPSALPAGRVIKPQVINQAVGLPASGSLYYFNYLDSTAYPPSELLSGHLFPSGLLNAQAPAALAAHPKLFLLENAKLAGLAADKFPHPAPYPHKERLPAPLEQVLKENSALTAERGGVKGHSKLPGGSADGKPKNFTCE | |
| 0.02 mL | |
| Neuroscience | |
| 55079 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction