missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ZNF313 Polyclonal Antibody

Product Code. 16500952
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16500952 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16500952 Supplier Invitrogen™ Supplier No. PA598121

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

ZNF313 Polyclonal Antibody for Western Blot, IHC (P), ELISA

ZNF313 is a novel human zinc finger protein gene that contains both ring finger and C(2)H(2) domain. It may play a role in spermatogenesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF313
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.19 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene RNF114
Gene Accession No. Q9ET26, Q9Y508
Gene Alias 1110008J21Rik; AI225886; AW549494; E3 ubiquitin-protein ligase RNF114; PSORS12; RING finger protein 114; RING-type E3 ubiquitin transferase RNF114; RNF114; Zfp228; Zfp313; Zinc finger protein 228; zinc finger protein 313; ZNF228; Znf313
Gene Symbols RNF114
Host Species Rabbit
Immunogen Recombinant Human E3 ubiquitin-protein ligase RNF114 protein (1-228AA).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55905, 81018
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.