missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF319 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-20989
This item is not returnable.
View return policy
Description
ZNF319 Polyclonal specifically detects ZNF319 in Human samples. It is validated for Western Blot.
Specifications
| ZNF319 | |
| Polyclonal | |
| Unconjugated | |
| 0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal | |
| Zinc finger and SCAN domain-containing protein 15, zinc finger protein 397, Zinc finger protein 47ZSCAN15MGC13250, ZNF47 | |
| Rabbit | |
| 66 kDa | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG |
| Western Blot | |
| 1mg/mL | |
| Western Blot 1:500-1:3000 | |
| Q9P2F9 | |
| ZNF319 | |
| Recombinant protein encompassing a sequence within the center region of human ZNF319. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| 57567 | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction