missing translation for 'onlineSavingsMsg'
Learn More

ZNF330 Monoclonal Antibody (OTI8E9), TrueMAB™, OriGene

Product Code. p-200015828
Encompass_Preferred
Change view
Click to view available options
Quantity:
30 μL
100 μL
Unit Size:
100µL
30µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17503014 30 μL 30µL
17583004 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17503014 Supplier Origene Technologies Supplier No. TA810145S

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

ZNF330 Monoclonal antibody specifically detects ZNF330 in Human samples. It is validated for Western Blot

Specifications

Antigen ZNF330
Applications Western Blot
Classification Monoclonal
Clone OTI8E9
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene SLC39A5
Gene Accession No. Q6ZMH5
Gene Alias BC008086; HSA6591; NOA 36; Noa36; Nucleolar autoantigen 36; nucleolar cysteine-rich protein; Zfp330; zinc finger 330-like protein; zinc finger autoantigen 330; zinc finger protein 330; zinc finger protein 330-like protein; Znf330
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 48-178 of human SLC39A5 produced in E.coli.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 283375
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.