missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ AMID Overexpression Lysate
Shop All Bio Techne Products
Click to view available options
Quantity:
0.1 mg
Conditionnement:
0.1mg
Description
The AMID Overexpression Lysate is derived from human. It has been validated for the following applications: Western Blot.
Spécification
Spécification
| Cell Line | The lysate was created in HEK293T cells, using plasmid ID RC24934 and based on accession number NM_32797. The protein contains a C-terminal DDK tag. |
| For Use With (Application) | Western Blot |
| Quantity | 0.1 mg |
| Storage Buffer | RIPA buffer |
| Purity | Protein |
| Test Specificity | Homo sapiens apoptosis-inducing factor, mitochondrion-associated, 2 (AIFM2), nuclear gene encoding mitochondrial protein, mRNA. |
| Content And Storage | -80°C. Avoid freeze-thaw cycles. |
| Protein State | Native |
For Research Use Only
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu