missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF34 Antibody (OTI3G2), Alexa Fluor™ 750, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74937AF750
This item is not returnable.
View return policy
Description
ZNF34 Monoclonal specifically detects ZNF34 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| ZNF34 | |
| Monoclonal | |
| Alexa Fluor 750 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| ZNF34 | |
| Recombinant protein fragment corresponding to amino acids 117-405 of human ZNF34 (NP_085057) produced in E.coli. | |
| Primary | |
| Human | |
| IgG1 |
| Western Blot, Immunohistochemistry | |
| OTI3G2 | |
| Western Blot, Immunohistochemistry | |
| FLJ3472, KOX21zinc finger protein 37a (KOX 21), zinc finger protein 37A, Zinc finger protein KOX21, ZNF37 | |
| Mouse | |
| 0.1 mL | |
| 80778 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction