Learn More
ZNF34 Monoclonal Antibody (OTI4E4), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA803568
This item is not returnable.
View return policy
Description
May be involved in transcriptional regulation.Specifications
| ZNF34 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q8IZ26 | |
| ZNF34 | |
| Human recombit protein fragment corresponding to amino acids 117-405 of human ZNF34 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI4E4 | |
| Unconjugated | |
| ZNF34 | |
| KOX32 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 80778 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |