missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF346 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-76540
This item is not returnable.
View return policy
Description
ZNF346 Polyclonal antibody specifically detects ZNF346 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| ZNF346 | |
| Polyclonal | |
| Unconjugated | |
| PBS | |
| KIAA0844ZNF365D, MGC41821, MGC87345, Su48, talanin, UANprotein ZNF365, zinc finger protein 365 | |
| Antibody was raised against a 20 amino acid synthetic peptide near the carboxy terminus of human ZNF346. The immunogen is located within the last 50 amino acids of ZNF346. Amino Acid Squence: LGQIPMQRQPIQKDSTTLED | |
| 0.1 mg | |
| Apoptosis | |
| 23567 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| Western Blot 1-2 μg/mL, ELISA 1:100-1:2000, Immunohistochemistry 5 μg/mL, Immunocytochemistry/ Immunofluorescence 20 μg/mL, Immunohistochemistry-Paraffin 5 μg/mL | |
| NP_036411 | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction