missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZNF346 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5118373
This item is not returnable.
View return policy
Description
ZNF346 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), IP
Probable transcriptional activator which may play a role in neuronal differentiation. Able to bind DNA and activate expression in vitro.
Specifications
| ZNF346 | |
| Polyclonal | |
| PBS with 0.03% ProClin 300; pH 7 | |
| Q9UL40 | |
| ZNF346 | |
| E. coli-derived Human ZNF346 fragment. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| ZNF346 | |
| double-stranded RNA-binding zinc finger protein JAZ; Jaz; Just another zinc finger protein; Zfp346; Zinc finger protein 346; ZNF346 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 23567 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction